Please login to your online account to display your discounted pricing

Alfa Aesar™ Decanonitrile, 98%

$47.32 - $650.92

Chemical Identifiers

Chemical Name or Material Decanonitrile
CAS 1975-78-6
Molecular Formula C10H19N
Linear Formula CH3(CH2)8CN
Formula Weight 153.27
MDL Number MFCD00039505
Synonym 1-Cyanononane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA187980B

  SDS
alfa aesar™
A187980B
1000g Each for $650.92

AAA1879830

  SDS
alfa aesar™
A1879830
250g Each for $183.82

AAA1879818

  SDS
alfa aesar™
A1879818
50g Each for $47.32
Description & Specifications

Specifications

Formula Weight 153.27
Chemical Name or Material Decanonitrile
TSCA TSCA
Assay 98%
Molecular Formula C10H19N
Linear Formula CH3(CH2)8CN
Boiling Point 241° to 243°C
MDL Number MFCD00039505
CAS 1975-78-6
Melting Point −15°C
Quantity 1000g
Synonym 1-Cyanononane
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Health Hazard 3 GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301
Toxic if swallowed.