missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Decanoic Anhydride, TCI America™

$119.17 - $366.67

Chemical Identifiers

CAS 2082-76-0
Molecular Formula C20H38O3
Molecular Weight (g/mol) 326.521
MDL Number MFCD00010460
InChI Key HTWWKYKIBSHDPC-UHFFFAOYSA-N
Synonym decanoic anhydride, capric anhydride, n-capric anhydride, n-decanoic anhydride, decanoic acid anhydride, acmc-209few, decanoic acid, anhydride, decanoic anhydride, decanoic acid,1,1'-anhydride, htwwkykibshdpc-uhfffaoysa
PubChem CID 74960
IUPAC Name decanoyl decanoate
SMILES CCCCCCCCCC(=O)OC(=O)CCCCCCCCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C002025G View Documents TCI America
C002025G
25g Each for $119.17
 
C0020100G View Documents TCI America
C0020100G
100g Each for $366.67
 
Specifications

Chemical Identifiers

2082-76-0
326.521
HTWWKYKIBSHDPC-UHFFFAOYSA-N
74960
CCCCCCCCCC(=O)OC(=O)CCCCCCCCC
C20H38O3
MFCD00010460
decanoic anhydride, capric anhydride, n-capric anhydride, n-decanoic anhydride, decanoic acid anhydride, acmc-209few, decanoic acid, anhydride, decanoic anhydride, decanoic acid,1,1'-anhydride, htwwkykibshdpc-uhfffaoysa
decanoyl decanoate

Specifications

White
2082-76-0
MFCD00010460
decanoic anhydride, capric anhydride, n-capric anhydride, n-decanoic anhydride, decanoic acid anhydride, acmc-209few, decanoic acid, anhydride, decanoic anhydride, decanoic acid,1,1'-anhydride, htwwkykibshdpc-uhfffaoysa
CCCCCCCCCC(=O)OC(=O)CCCCCCCCC
326.521
326.52
≥98.0% (GC)
25g
C20H38O3
3261
HTWWKYKIBSHDPC-UHFFFAOYSA-N
decanoyl decanoate
74960
Crystal-Lump at 20°C
Decanoic Anhydride
Safety and Handling

Safety and Handling

EINECSNumber : (2)-3916

TSCA : No

Recommended Storage : Refrigerator