missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Decane, 99%, anhydrous, AcroSeal™, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC434600010

 View more versions of this product

Catalog No. AC434600010



Specifications

Specifications

Decane
46°C
AcroSeal™ Glass Bottle
0.0005 % max.
0.005% Max.
Colorless
1L
98.5% min. (GC)
CH3(CH2)8CH3
Solubility in water: insoluble. Other solubilities: soluble in alcohol (miscible with ethanol),soluble in ether,slightly soluble in ccl4
CCCCCCCCCC
142.28
CHEBI:41808
Liquid
0.7350g/mL
Authentic
1.4103 to 1.4123
0.735
174.0°C
-30.0°C
124-18-5
C10H22
ccris 653, decane, analytical standard, decyl hydride, dsstox_cid_4913, dsstox_gsid_24913, dsstox_rid_77577, hsdb 63, n-decane, nonane, methyl, unii-nk85062oiy
DIOQZVSQGTUSAI-UHFFFAOYSA-N
decane
15600
142.28
99%

Chemical Identifiers

124-18-5
142.28
ccris 653, decane, analytical standard, decyl hydride, dsstox_cid_4913, dsstox_gsid_24913, dsstox_rid_77577, hsdb 63, n-decane, nonane, methyl, unii-nk85062oiy
CHEBI:41808
CCCCCCCCCC
C10H22
DIOQZVSQGTUSAI-UHFFFAOYSA-N
15600
decane
Safety and Handling

Safety and Handling

GHS H Statement
May be fatal if swallowed and enters airways.
Flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.

GHS Signal Word: Danger

EINECSNumber : 204-686-4

SDS Documents