Learn More
Decanal, 96%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A11656AP
Description
Decanal is used in fragrances and flavoring and it is an important component in citrus along with octanal, citral, and sinensal. It is also used in Air Care Products, Laundry and Dishwashing Products, Personal Care Products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDecanal is used in fragrances and flavoring and it is an important component in citrus along with octanal, citral, and sinensal. It is also used in Air Care Products, Laundry and Dishwashing Products, Personal Care Products.
Solubility
Insoluble in water.
Notes
Air Sensitive. Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Decanal | |
-5°C | |
207°C to 209°C | |
96% | |
1.428 | |
MFCD00007031 | |
1362530 | |
decyl aldehyde, capraldehyde, caprinaldehyde, decanaldehyde, n-decyl aldehyde, capric aldehyde, 1-decanal, n-decanal, n-decaldehyde, 1-decyl aldehyde | |
KSMVZQYAVGTKIV-UHFFFAOYSA-N | |
decanal | |
8175 | |
156.27 | |
Liquid |
112-31-2 | |
0.828 | |
85°C (185°F) | |
C10H20O | |
CH3(CH2)8CHO | |
500 mL | |
Air Sensitive | |
Insoluble in water. | |
CCCCCCCCCC=O | |
156.269 | |
CHEBI:31457 | |
96% |
Chemical Identifiers
112-31-2 | |
156.269 | |
KSMVZQYAVGTKIV-UHFFFAOYSA-N | |
8175 | |
decanal |
C10H20O | |
MFCD00007031 | |
decyl aldehyde, capraldehyde, caprinaldehyde, decanaldehyde, n-decyl aldehyde, capric aldehyde, 1-decanal, n-decanal, n-decaldehyde, 1-decyl aldehyde | |
CHEBI:31457 | |
CCCCCCCCCC=O |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P374-P501c
H227-H315-H319-H335
EINECSNumber : 203-957-4
RTECSNumber : HD6000000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only