missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Decamethylcyclopentasiloxane, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 454580500
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Decamethylcyclopentasiloxane | |
0.9590g/mL | |
76°C | |
C10H30O5Si5 | |
0.959 | |
decamethylcyclopentasiloxane, cyclopentasiloxane, decamethyl, cyclomethicone 5, dimethylsiloxane pentamer, dow corning 345 fluid, nuc silicone vs 7158, dow corning 345, silicon sf 1202, cyclic dimethylsiloxane pentamer, dekamethylcyklopentasiloxan | |
XMSXQFUHVRWGNA-UHFFFAOYSA-N | |
2,2,4,4,6,6,8,8,10,10-decamethyl-1,3,5,7,9,2,4,6,8,10-pentaoxapentasilecane | |
10913 | |
370.77 |
541-02-6 | |
101.0°C (20.0 mmHg) | |
96% min. (GC) | |
50g | |
15,2852 | |
Solubility in water: insoluble | |
C[Si]1(O[Si](O[Si](O[Si](O[Si](O1)(C)C)(C)C)(C)C)(C)C)C | |
370.77 | |
4 mPa.s (20°C) | |
97% |
Safety and Handling
EINECSNumber : 208-764-9
RUO â Research Use Only