Please login to your online account to display your discounted pricing

Decahydronaphthalene (Laboratory), Fisher Chemical


Quantity 500mL
Packaging Glass Bottle

Manufacturer: fisher scientific company  BPD0500

 View more versions of this product

Catalog No. D0-500

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Laboratory Grade
Boiling Point 187°C
Formula Weight 138.25
Melting Point −31°C
Quantity 500mL
DOT Information DOT Class 3, : Flammable Liquid
Physical Form Liquid
Molecular Formula C10H18
Vapor Pressure 1.3mbar at 22°C
Color Colorless
pH 7
Density 0.890g/cm³
UN Number 1147
Health Hazard 2 WARNING!
Chemical Name or Material Decahydronaphthalene
Packaging Glass Bottle
Additional Information Vapor Pressure: 1.3mbar at 22°C
MDL Number MFCD00004130
CAS 91-17-8
ChemAlert Storage Symbol Red
Synonym Decalin
Health Hazard 3 Emergency Overview
Flammable liquid and vapor. Harmful by inhalation. Causes eye, skin, and respiratory tract irritation. Aspiration hazard if swallowed - can enter lungs and cause damage. Hygroscopic. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. If peroxide formation is suspected, do not open or move container. Use only under a chemical fume hood. Handle under an inert atmosphere. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes.

NFPA

Health:2
Flammability:2
Instability:1
Identification Pass Test

Specifications

Chemical Name or Material Decahydronaphthalene
CAS 91-17-8
Purity Grade Laboratory Grade
Molecular Formula C10H18
Formula Weight 138.25
MDL Number MFCD00004130
Synonym Decalin
Accessories
 

Emergency Overview
Flammable liquid and vapor. Harmful by inhalation. Causes eye, skin, and respiratory tract irritation. Aspiration hazard if swallowed - can enter lungs and cause damage. Hygroscopic. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. If peroxide formation is suspected, do not open or move container. Use only under a chemical fume hood. Handle under an inert atmosphere. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes.

NFPA

Health:2
Flammability:2
Instability:1

WARNING!

SDS & Safety Documents