missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Decahydro-2-naphthol, 98%, mixture of cis and trans, ACROS Organics™

Chemical Identifiers

CAS 825-51-4
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.253
MDL Number MFCD00004132
InChI Key UPMAOXLCTXPPAG-UHFFFAOYSA-N
Synonym 2-decalinol, 2-decalol, 2-hydroxydecalin, 2-naphthalenol, decahydro, 2-naphthol, decahydro, decahydro-2-naphthol, decahydronaphthalen-2-ol, decahydronaphthol-2, decahydronapthol-2, decalin-2-ol
PubChem CID 13216
IUPAC Name 1,2,3,4,4a,5,6,7,8,8a-decahydronaphthalen-2-ol
SMILES C1CCC2CC(CCC2C1)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC111851000
SDS View Documents
Thermo Fisher Scientific
111851000
100g Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

825-51-4
154.253
UPMAOXLCTXPPAG-UHFFFAOYSA-N
13216
C1CCC2CC(CCC2C1)O
C10H18O
MFCD00004132
2-decalinol, 2-decalol, 2-hydroxydecalin, 2-naphthalenol, decahydro, 2-naphthol, decahydro, decahydro-2-naphthol, decahydronaphthalen-2-ol, decahydronaphthol-2, decahydronapthol-2, decalin-2-ol
1,2,3,4,4a,5,6,7,8,8a-decahydronaphthalen-2-ol

Specifications

Authentic
White to Yellow
825-51-4
C10H18O
MFCD00004132
2-decalinol, 2-decalol, 2-hydroxydecalin, 2-naphthalenol, decahydro, 2-naphthol, decahydro, decahydro-2-naphthol, decahydronaphthalen-2-ol, decahydronaphthol-2, decahydronapthol-2, decalin-2-ol
C1CCC2CC(CCC2C1)O
154.253
154.25
≥97.5%
Glass Bottle
100g
98%
C10H17OH
06, 67
UPMAOXLCTXPPAG-UHFFFAOYSA-N
1,2,3,4,4a,5,6,7,8,8a-decahydronaphthalen-2-ol
13216
Weak Paste
Decahydro-2-naphthol, 98%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 212-545-3

RTECSNumber : QK4240000

TSCA : TSCA

SDS
Product Certifications