Please login to your online account to display your discounted pricing

Alfa Aesar™ Decafluorobiphenyl, 99%

$84.04 - $287.85

Chemical Identifiers

Chemical Name or Material Decafluorobiphenyl
CAS 434-90-2
Molecular Formula C12F10
Formula Weight 334.12
MDL Number MFCD00000292
Beilstein 1916978
Synonym Perfluorobiphenyl
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1594806

  SDS
alfa aesar™
A1594806
5g Each for $84.04

AAA1594814

  SDS
alfa aesar™
A1594814
25g Each for $287.85
Description & Specifications

Specifications

Formula Weight 334.12
Chemical Name or Material Decafluorobiphenyl
Assay 99%
Molecular Formula C12F10
Boiling Point 206°C
MDL Number MFCD00000292
CAS 434-90-2
Melting Point 66° to 70°C
Beilstein 1916978
Quantity 5g
Synonym Perfluorobiphenyl

Applications
Decafluorobiphenyl is used as an intermediate in organic synthesis and in pharmaceuticals.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.