Please login to your online account to display your discounted pricing

Alfa Aesar™ Decafluorobenzophenone, 98%

$56.96 - $217.04

Chemical Identifiers

Chemical Name or Material Decafluorobenzophenone
CAS 853-39-4
Molecular Formula C13F10O
Formula Weight 362.12
MDL Number MFCD00000295
Beilstein 2066476
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1926203

 
alfa aesar™
A1926203
1g Each for $56.96

AAA1926206

 
alfa aesar™
A1926206
5g Each for $217.04
Description & Specifications

Specifications

Formula Weight 362.12
Beilstein 2066476
Chemical Name or Material Decafluorobenzophenone
Assay 98%
Molecular Formula C13F10O
MDL Number MFCD00000295
CAS 853-39-4
Melting Point 92° to 94°C
Quantity 1g

Applications
Perfluorobenzophenone was used in the synthesis of fluorinated acridones and acridines. Fluorinated polyetherketones prepared from decafluorobenzophenone. Laser pulse excitation of decafluorobenzophenone (DFB) in acetonitrile yields the triplet 3DFB.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.