Please login to your online account to display your discounted pricing

Alfa Aesar™ Decafluorobenzhydrol, 97%

$122.69 - $122.69

Chemical Identifiers

Chemical Name or Material Decafluorobenzhydrol
CAS 1766-76-3
Molecular Formula C13H2F10O
Formula Weight 364.14
MDL Number MFCD00000297
Beilstein 2067248
Synonym Bis(pentafluorophenyl) carbinol; Bis(pentafluorophenyl)methanol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0219706

 
alfa aesar™
L0219706
5g Each for $122.69
Description & Specifications

Specifications

Formula Weight 364.14
Chemical Name or Material Decafluorobenzhydrol
Assay 97%
Molecular Formula C13H2F10O
MDL Number MFCD00000297
CAS 1766-76-3
Melting Point 77° to 80°C
Beilstein 2067248
Quantity 5g
Synonym Bis(pentafluorophenyl) carbinol; Bis(pentafluorophenyl)methanol
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.