Learn More
Decacarbonyldirhenium, 96%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 06210803
Description
Decacarbonyldirhenium is a key starting material used for the synthesis of other rhenium carbonyl complexes. It plays an important role in metathesis, reforming and hydrogenation reactions. It is used as a catalyst to promote silation of alcohols and in the preparion of silyl ethers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Decacarbonyldirhenium | |
170°C (decomposition) | |
2.87 g/mL | |
Re2(CO)10 | |
1 g | |
carbon monoxide; rhenium 2+, decacarbonyldirhenium 250mg | |
[O]#C[Re](C#[O])(C#[O])(C#[O])(C#[O])[Re](C#[O])(C#[O])(C#[O])(C#[O])C#[O] | |
652.51 | |
652.52 | |
Powder |
14285-68-8 | |
White | |
C10O10Re2 | |
MFCD00011198 | |
UN3466 | |
ZIZHEHXAMPQGEK-UHFFFAOYSA-N | |
carbon monoxide;rhenium | |
498777 | |
96% |
Chemical Identifiers
14285-68-8 | |
652.51 | |
ZIZHEHXAMPQGEK-UHFFFAOYSA-N | |
498777 |
C10O10Re2 | |
MFCD00011198 | |
carbon monoxide; rhenium 2+, decacarbonyldirhenium 250mg | |
[O]#C[Re](C#[O])(C#[O])(C#[O])(C#[O])[Re](C#[O])(C#[O])(C#[O])(C#[O])C#[O] |
Safety and Handling
GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.
P261-P271-P304+P340-P312
H332
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: METAL CARBONYLS, SOLID, N.O.S.
EINECSNumber : 238-202-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only