Please login to your online account to display your discounted pricing

Alfa Aesar™ Decacarbonyldimanganese, C 30.6%

Chemical Identifiers

Chemical Name or Material Decacarbonyldimanganese
CAS 10170-69-1
Linear Formula Mn2(CO)10
Formula Weight 389.98
MDL Number MFCD00011115
UN Number UN3466
Merck Index 14,5727
Synonym Manganese carbonyl; Dimanganese decacarbonyl
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1305803

  SDS
alfa aesar™
1305803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1305806

  SDS
alfa aesar™
1305806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1305814

  SDS
alfa aesar™
1305814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 389.98
Health Hazard 3 GHS P Statement
P280f-P304+P340-P309-P310
Wear protective clothing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
TSCA TSCA
Physical Form Crystalline
Linear Formula Mn2(CO)10
Color Yellow
CAS 10170-69-1
Melting Point 152° to 154°C
Merck Index 14,5727
Quantity 1g
Synonym Manganese carbonyl; Dimanganese decacarbonyl
Solubility Information Soluble in organic solvents; Insoluble in water
Health Hazard 1 Danger
Density 1.75g/mL
Health Hazard 2 GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.
Chemical Name or Material Decacarbonyldimanganese
Boiling Point 80°C (1mmHg) (subl.)
MDL Number MFCD00011115
UN Number UN3466
Concentration C 30.6%
Sensitivity Air Sensitive
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: METAL CARBONYLS, SOLID, N.O.S.

  • Starting material for manganese carbonyl chemistry, as a catalyst and an antiknock additive

Safety and Handling

Danger

GHS P Statement
P280f-P304+P340-P309-P310
Wear protective clothing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.