Please login to your online account to display your discounted pricing

Alfa Aesar™ Decaborane, 98%

In rocket propellants, and as a catalyst in olefin polymerization

Chemical Identifiers

Chemical Name or Material Decaborane
CAS 17702-41-9
Molecular Formula B10H14
Formula Weight 122.22
MDL Number MFCD00011314
UN Number UN1868
Merck Index 14,2845
Synonym Decaboron tetradecahydride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA8789203

  SDS
alfa aesar™
8789203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA8789206

  SDS
alfa aesar™
8789206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA8789214

 
alfa aesar™
8789214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 122.22
Chemical Name or Material Decaborane
TSCA TSCA
Physical Form Powder
Assay 98%
Molecular Formula B10H14
Boiling Point 213°C
MDL Number MFCD00011314
Color White to pale yellow
CAS 17702-41-9
UN Number UN1868
Melting Point 99° to 103°C
Merck Index 14,2845
Quantity 1g
Synonym Decaboron tetradecahydride
Odor Pungent
Solubility Information Slightly soluble in cold water; Hydrolyzes in hot water; Soluble in ethyl acetate, 1-Bromopropane, ethyl silicate, benzene, alcohol, and carbon tetrachloride
Health Hazard 1 Danger
Density 0.940g/mL
Health Hazard 2 GHS H Statement
H228-H301-H310-H330
Flammable solid.
Toxic if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Health Hazard 3 GHS P Statement
P210-P301+P310-P304+P340-P320-P330-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: DECABORANE
RTECS Number HD1400000
Safety and Handling

Danger

GHS P Statement
P210-P301+P310-P304+P340-P320-P330-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H228-H301-H310-H330
Flammable solid.
Toxic if swallowed.
Fatal in contact with skin.
Fatal if inhaled.