missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Deblock trichloroacetic acid soln., 3% w/w in dichloromethane

Specifications

Chemical Name or Material Deblock trichloroacetic acid soln.
MDL Number MFCD00004177
UN Number UN2810
Synonym DEBLK soln.
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ62821AH View Documents Alfa Aesar™
J62821AH
200mL N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAJ62821K4 View Documents Alfa Aesar™
J62821K4
2.5L N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAJ62821AO View Documents Alfa Aesar™
J62821AO
450mL N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Solvent specially purified and analyzed for biotechnology applications, including peptide and oligonucleotide synthesis.
Specifications

Specifications

Deblock trichloroacetic acid soln.
MFCD00004177
UN2810
DEBLK soln.