Learn More
Dealing with Murder Forensic Activity Series
Combination CD with PDFs leads teacher and students through murder and mayhem.
$127.10 - $127.10
Specifications
For Use With (Application) | For directions and instructions support students and teachers as they work through the investigation |
---|---|
Includes | Pdf's of a crime file, crime log, suspect chart and crime scene sketch, as well as each day's evidence and clues |
Includes: Each activity includes PDFs of a crime file, crime log, suspect chart and crime scene sketch, as well as each day's evidence and clues. Directions and instructions support students and teachers as they work through the investigation.
Catalog Number | Mfr. No. | Product Type | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity | |||||
S99284B
|
Fisher Science Education
FE10 |
Dealing With Murder Forensic Activity: Fatal Error |
Each for $127.10
|
|
Description
Merging science and literacy, students are required to be critical and active readers as they conduct their own investigation. Teacher-created activity facilitates the execution of NSES scientific literacy standards.
Students begin with an introduction to the crime and evaluation of the crime scene photos, and then investigate, extract and analyze key information; formulate hypotheses; deduce the murderer's identity from among a number of interesting suspects and finally, recreate the murder event and finger the murderer.
- Created by teachers for teachers with students in grades 7 to 12
- Activity facilitates the execution of the scientific literacy standards as defined by the National Science Education Standards
Specifications
For directions and instructions support students and teachers as they work through the investigation | |
Pdf's of a crime file, crime log, suspect chart and crime scene sketch, as well as each day's evidence and clues |