missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Mopec DE300 Roller Autopsy Cart
Gliding on corrosion proof nylon rollers, the body tray easily rolls onto the cart frame. Mopec™ DE300 Roller Autopsy Cart is designed with a built in slope and can be used in conjunction with any Mopec™ wall mounted sinks.
Supplier: Mopec DE300
Description
- All stainless steel construction with heavy duty 1.5 inch stainless steel tubing frame
- Heavy duty 8 in. casters with total lock feature
- Corrosion proof nylon rollers
- Compatible with Mopec™ autopsy sinks
Warranty
1 Year Warranty
Specifications
Autopsy Cart | |
31 in. | |
27.25 in. | |
200.66 cm |
Stainless Steel | |
79 in. | |
78.74 cm | |
69.21 cm |