Invitrogen™ DDR2 Recombinant Human Protein DFS Item
DDR2 is a member of a subclass of RTKs and contains a distinct extracellular region encompassing a factor VIII-like domain.
Manufacturer:  Invitrogen™ LSPV4188
 View more versions of this product
Catalog No. PV4188
Description
Description
DDR1 and DDR2 have been shown to be potently inhibited by Gleevec.
Specifications
Specifications
NP_006173, S642A
Frozen
79.8kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Cytoplasmic
Kinase Assay
DDR Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
DDR2, TK, NTRKR3
Baculovirus
Human
GST Tag
Documents