Please login to your online account to display your discounted pricing

DCG IV, Tocris


Very potent, selective group II mGlu agonist. Also NMDA agonist

Quantity 10mg

Manufacturer: r & d systems  RDS097510

 View more versions of this product

Catalog No. 097510

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material DCG IV
CAS 147782-19-2
Recommended Storage Store at -20°C
Assay Percent Range >98%
Molecular Formula C7H9NO6
Formula Weight 203.15
Synonym (2S,2'R,3'R)-2-(2',3'-Dicarboxycyclopropyl)glycine
Quantity 10mg

Specifications

Chemical Name or Material DCG IV
Assay Percent Range >98%
Molecular Formula C7H9NO6
Formula Weight 203.15
Synonym (2S,2'R,3'R)-2-(2',3'-Dicarboxycyclopropyl)glycine
Accessories

Store at -20°C