missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DCG IV, Tocris

Very potent, selective group II mGlu agonist. Also NMDA agonist

$554.62 - $2,331.24

Chemical Identifiers

Assay Percent Range >98%
Molecular Formula C7H9NO6
Formula Weight 203.15
Synonym (2S,2'R,3'R)-2-(2',3'-Dicarboxycyclopropyl)glycine
Chemical Name or Material DCG IV
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
097550 R & D Systems
RDS097550
50mg Each for $2,331.24
097510 R & D Systems
RDS097510
10mg Each for $554.62
Specifications Description & Specifications

Chemical Identifiers

>98%
203.15
DCG IV
C7H9NO6
(2S,2'R,3'R)-2-(2',3'-Dicarboxycyclopropyl)glycine

Specifications

147782-19-2
C7H9NO6
(2S,2'R,3'R)-2-(2',3'-Dicarboxycyclopropyl)glycine
DCG IV
>98%
203.15
50mg

Safety and Handling

Recommended Storage : Store at -20°C