Please login to your online account to display your discounted pricing

DCB, Tocris

Allosteric modulator at mGlu5

$129.05 - $546.19

Chemical Identifiers

Chemical Name or Material DCB
Assay Percent Range >98%
Molecular Formula C14H10Cl2N2
Formula Weight 277.15
Synonym [(3-Chlorophenyl)methylene]hydrazone-3-chlorobenzaldehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

195250

 
r & d systems
RDS195250
50mg Each for $546.19

195210

 
r & d systems
RDS195210
10mg Each for $129.05
Description & Specifications

Specifications

Formula Weight 277.15
Quantity 50mg
Chemical Name or Material DCB
Assay Percent Range >98%
Molecular Formula C14H10Cl2N2
Synonym [(3-Chlorophenyl)methylene]hydrazone-3-chlorobenzaldehyde
CAS 6971-97-7
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature