Learn More
3M™ DBI-SALA™ Vest-Type Harnesses
Description
- No-tangle design
- Stand-up back D-ring
- Repel™ webbing
- Built-in lanyard keepers
- Revolver™ vertical torso adjusters
- Impact Indicator
Order Info
One size fits most workers. Choice of leg strap buckles.
Compliance
Meet ANSI™ Z359.1-1992, ANSI A10.14-1991, and OSHA requirements.
Specifications
Specifications
| Material | Polyester |
| Certifications/Compliance | Meets ANSI Z359.1-1992, ANSIFor Use With (Application) A10.14-1991, OSHA |
| Size | Universal |
| D-Ring Location | Back |
| Style | Vest |
| Buckle Style | Friction buckle |