Please login to your online account to display your discounted pricing
DBI/Sala™ Sealed Self-Retracting Lifeline
Removes the danger of trailing lines
$1,943.22 - $1,943.22
Specifications
Certifications/Compliance ANSI Z359.1-1992, ANSI A10.14-1991, OSHA
Product Type Self retracting lifeline
View More Specs
Products
Catalog Number Mfr. No. Includes Material Length (English) Length (Metric) Price Quantity    
19-809-546
capital safety group cnd
3403401
Stainless Steel cable Stainless Steel 50 ft. 15.24m Each for $1,943.22
Description & Specifications
Specifications
Certifications/Compliance ANSI Z359.1-1992, ANSI A10.14-1991, OSHA
Product Type Self retracting lifeline
Motor spring and brake components are contained inside a sealed drum, protecting them from dirt, dust, moisture, and other elements, and increasing reliability.
  • Spring-tensioned 3/16 in. (5mm) cable automatically retracts as worker moves closer to the anchor point
  • If a worker slips or falls, the braking mechanism stops the fall within approximately 18 in. (45.7cm)
  • Limits the forces imposed on the worker to 900 lb. (409kg) or less
  • Molded urethane bumper protects lifeline from kinks
  • Reposition bumper to allow inspection
  • Contains reserve lifeline system and impact-indicating swiveling snap hook
  • Cast-aluminum housing, stainless-steel housing cover, mainshaft, fasteners, and locking pawls
Maximum workload: 310 lb. (141kg). Average locking speed: 4 1/2 in. (11.4cm) per second. Optional carrying cases available; see cat. No. 19-042-008.
Meets ANSI Z359.1-1992, ANSI A10.14-1991, and OSHA requirements.
Sealed drum assembly was fully functional following consecutive corrosion tests that included a 100-hour salt spray per ASTM B 117-90 and a 10-day humidity cycle per MIL-STD-810.