missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3M™ DBI-SALA™ Mounted Recovery System: Winches

$2,806.05 - $2,806.05

Specifications

Certifications/Compliance OSHA 1910.28, 1926.451, OSHA 1910.146, ANSI Z117.1
Color Silver
Product Type Salalift Winch
For Use With (Equipment) Mounted recovery system
View More Specs

Products 1
Catalog Number Mfr. No. Includes Length (English) Material Weight (English) Price Quantity  
Catalog Number Mfr. No. Includes Length (English) Material Weight (English) Price Quantity  
19-057006
View Documents
3M™
8101000
60 ft. galvanized steel cable, mounting bracket, triple braking system, quick mount bracket, carrying bag 60 ft. 0.25 in. galvanized steel cable 62 lb.
Each for $2,806.05
 
Description

Description

Designed for raising, lowering, work support or personnel rescue

  • Triple braking system includes two independent centrifugal brakes and one dead-man type brake for added security
  • Two-speed input (primary 6.6:2, secondary 3.3:1) for versatility and ease of use
  • Durable corrosion-resistant construction featuring a heat-treated cast aluminum housing with steel gears contained in a sealed oil bath
  • Overload clutch (load limiting input handle) reduces chance of injury to entangled worker
  • Complete with unique quick-mount bracket making attachment to DBI/SALA support structures simple and easy
  • Underwriters Laboratories (UL) tested and classified
  • Meets or exceeds all applicable industry standards including OSHA 1910.28 and 1926.451, OSHA 1910.146 and ANSI Z117.1

Specifications

Specifications

OSHA 1910.28, 1926.451, OSHA 1910.146, ANSI Z117.1
Silver
Salalift Winch
Mounted recovery system
SDS