3M™ DBI-SALA™ Mounted Recovery System: Winches
Manufacturer:  3M™ 8101000
 View more versions of this product
Catalog No. 19-057006
Description
Description
Designed for raising, lowering, work support or personnel rescue
  • Triple braking system includes two independent centrifugal brakes and one dead-man type brake for added security
  • Two-speed input (primary 6.6:2, secondary 3.3:1) for versatility and ease of use
  • Durable corrosion-resistant construction featuring a heat-treated cast aluminum housing with steel gears contained in a sealed oil bath
  • Overload clutch (load limiting input handle) reduces chance of injury to entangled worker
  • Complete with unique quick-mount bracket making attachment to DBI/SALA support structures simple and easy
  • Underwriters Laboratories (UL) tested and classified
  • Meets or exceeds all applicable industry standards including OSHA 1910.28 and 1926.451, OSHA 1910.146 and ANSI Z117.1
Specifications
Specifications
OSHA 1910.28, 1926.451, OSHA 1910.146, ANSI Z117.1
60 ft. galvanized steel cable, mounting bracket, triple braking system, quick mount bracket, carrying bag
Silver
0.25 in. galvanized steel cable
Mounted recovery system
62 lb.
60 ft.
Salalift Winch