Please login to your online account to display your discounted pricing
DBI/Sala™ Mounted Recovery System—Complete System
$7,236.70 - $7,236.70
Specifications
Certifications/Compliance OSHA 1910.28, 1926.451, OSHA 1910.146, ANSI Z117.1
For Use With (Application) Ideal for confined space rescue, work support, fall arrest
Load Bearing Capacity (English) 310 lb.
Load Bearing Capacity (Metric) 141kg
View More Specs
Products
Catalog Number Mfr. No. Includes Strength Price Quantity    
29-635-1823
capital safety group cnd
8304001
Sealed self retracting lifeline, adjustable Aluminum tripod, salalift winch and carrying bags, lifeline and winches are Stainless Steel 3000, 3600 (cable) lbf. Each for $7,236.70
Description & Specifications
Specifications
Certifications/Compliance OSHA 1910.28, 1926.451, OSHA 1910.146, ANSI Z117.1
For Use With (Application) Ideal for confined space rescue, work support, fall arrest
Load Bearing Capacity (English) 310 lb.
Load Bearing Capacity (Metric) 141kg