3M™ DBI-Sala™ ExoFit™ Vest-Style Harness
No-tangle “X” design dons easily
Manufacturer:  3M™ 1107977C
 View more versions of this product
Catalog No. 19-066-862C
Description
Description
Offering exceptional comfort and protection, the ExoFit™ features a no-tangle “X” design with memory fit, so it dons easily and only has to be adjusted once.
  • Made of soft, lightweight, high-performance materials for all-day wearability
  • Features built-in shoulder, hip, and leg padding with Lycra™ edging for chafe-free mobility
  • Aero-Spacer™ Dri-Lex™ lining wicks moisture away from the body for cool, dry comfort
  • Three quick-connect buckles facilitate rapid donning and adjustment even with just one hand
  • With back D-ring for connection to fall arrest system; convenient built-in belt; and coated, corrosion-resistant hardware
Specifications
Specifications
Meets all applicable OSHA, ANSI, and CSA requirements, including ANSIFor Use With (Application) Z359.
Back
Large