3M™ DBI-Sala™ Construction Cross-over Style Harnesses
Construction harness with parachute and pass-through buckles
Manufacturer: 3M™ 1101810C
Description
- Material: Polyester
- Cross-over style
- No-tangle functionality
- Front and back D-rings
- Tongue buckle body belt w/foam back pad
- Pass through legs
- Belt and hip pad with side D-rings
- Available in a variety of sizes
Specifications
Back, Side | |
Medium | |
Cross-over | |
Polyester webbing |