Please login to your online account to display your discounted pricing

DAU 5884 hydrochloride, Tocris

M3 receptor antagonist

$188.60 - $794.96

Chemical Identifiers

Chemical Name or Material DAU 5884 hydrochloride
Assay Percent Range >99%
Molecular Formula C17H21N3O3·HCl
Formula Weight 351.83
Synonym 8-Methyl-8-azabicyclo-3-endo[3.2.1]oct-3-yl-1, 4-dihydro-2-oxo-3(2H)-quinazolinecarboxylic acid ester hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

209610

 
r & d systems
209610
10mg Each for $188.60

209650

 
r & d systems
209650
50mg Each for $794.96
Description & Specifications

Specifications

Formula Weight 351.83
Quantity 10mg
Chemical Name or Material DAU 5884 hydrochloride
Molecular Formula C17H21N3O3·HCl
Synonym 8-Methyl-8-azabicyclo-3-endo[3.2.1]oct-3-yl-1, 4-dihydro-2-oxo-3(2H)-quinazolinecarboxylic acid ester hydrochloride
CAS 131780-48-8
Assay Percent Range >99%
Recommended Storage Desiccate at +4°C
Safety and Handling

Desiccate at +4°C