Please login to your online account to display your discounted pricing

Dapoxetine hydrochloride, Tocris™

Selective serotonin reuptake inhibitor

$139.08 - $585.69

Chemical Identifiers

Chemical Name or Material Dapoxetine hydrochloride
Assay Percent Range >99%
Molecular Formula C21H23NO·HCl
Formula Weight 341.87
Synonym (S)-N, N-Dimethyl-α-[2-(1-naphthalenyloxy)ethyl]benzenemethanamine hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

435810

  SDS
r & d systems
RDS435810
10mg Each for $139.08

435850

 
r & d systems
RDS435850
50mg Each for $585.69
Description & Specifications

Specifications

Formula Weight 341.87
Quantity 10mg
Chemical Name or Material Dapoxetine hydrochloride
Recommended Storage Desiccate at Room Temperature
Synonym (S)-N, N-Dimethyl-α-[2-(1-naphthalenyloxy)ethyl]benzenemethanamine hydrochloride
CAS 129938-20-1
Assay Percent Range >99%
Molecular Formula C21H23NO·HCl
Safety and Handling

Desiccate at Room Temperature