Invitrogen™ DAPK3 (ZIPK) Recombinant Human Protein DFS Item
DAPK3 is a serine/threonine protein kinase.
Manufacturer:  Invitrogen™ LSPV3995
 View more versions of this product
Catalog No. PV3995
Description
Description
DAPK3 is functionally linked to cell motility, smooth muscle contractility and cell death processes.
Specifications
Specifications
NP_001339
Frozen
80kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
Kinase Assay
DAPK Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
DAPK3 (ZIPK)
Baculovirus
Human
GST Tag
Documents