missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ DAPK2 Recombinant Human Protein
DAPK2 is a calcium/calmodulin-dependent kinase.
Supplier: Invitrogen™ PV3994
Description
DAPK2 acts as a positive regulator of apoptosis and is expressed in many tissue types.
Specifications
NP_055141 | |
50mM Tris-HCl, pH 7.5, 300mM NaCl, 0.5mM EDTA, 0.05% Brij-35, 2mM DTT, 50% glycerol | |
1mg | |
RUO - research use only | |
DAPK2, DRP-1 | |
Human | |
His Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
47.9kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
DAPK Kinase Family | |
Recombinant | |
Baculovirus |