missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DAPK2 Recombinant Human Protein
DAPK2 Recombinant Human Protein
Manufacturer: Life Technologies LSPV3614
Description
DAPK2 is a calcium⁄calmodulin-dependent kinase that acts as a positive regulator of apoptosis, and is expressed in many tissue types.Specifications
NP_055141 | |
Cell viability, proliferation and function, pharma and biopharma, protein assays and analysis, target and lead identification and validation | |
47.9kDa | |
85% by SDS-PAGE | |
–80°C | |
1 Each | |
DAPK2, DRP-1 | |
DAPK2 was pre-diluted in enzyme dilution buffer and assayed in 12.5mM Tris (pH 7.5), 10mM MgCl2, 1mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 4.8μM calmodulin, 0.5mM CaCl2, 2.5mM DTT, 0.01% Triton™ X-100, 200μM ATP, 167μg/mL MLCK peptide substrate (KKRAARATSDVFA-NH2) and trace [32P]-γ-ATP for 10 minutes at 30°C | |
Kinase and inhibitors | |
DAPK | |
18nmoles of phosphate transferred to MLCK peptide substrate (KKRAARATSDVFA-NH2) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 8.33μg/mL | |
His Tag |
0.34mg/mL | |
23604 | |
DAPK2 Recombinant Human Protein | |
10μg | |
For research use only. Not for use in diagnostic procedures | |
Serine/threonine kinase | |
DAPK2 Recombinant Human Protein | |
Kinase assay | |
Baculovirus | |
DAPK2 Recombinant Human Protein | |
Human |
For Research Use Only. Not for use in diagnostic procedures.