Please login to your online account to display your discounted pricing

Dansylamide, 99%, ACROS Organics™

$75.89 - $75.89

Chemical Identifiers

Chemical Name or Material Dansylamide
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C12H14N2O2S
Linear Formula (CH3)2NC10H6SO2NH2
Formula Weight 250.31
MDL Number MFCD00004000
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC204090010

  SDS
fisher scientific acros
204090010
Glass bottle 1g Each for $75.89
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Fine Crystalline Powder
Formula Weight 250.31
Quantity 1g
Color White to light yellow
Beilstein 14, IV, 2801
Melting Point 217° to 221°C
CAS 1431-39-6
Chemical Name or Material Dansylamide
Packaging Glass bottle
Assay Percent Range 99%
Molecular Formula C12H14N2O2S
Linear Formula (CH3)2NC10H6SO2NH2
MDL Number MFCD00004000
Infrared Spectrum Authentic

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.