Please login to your online account to display your discounted pricing

Alfa Aesar™ Dansyl-Tyr-Val-Gly

For cellular and molecular biology applications

$68.35 - $200.11

Specifications

Product Type Dansyl-Tyr-Val-Gly
Molecular Weight 570.66
Storage Requirements -30° to -10°C
Sequence Dansyl-Y-V-G
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66399LB0

 
alfa aesar™
J66399LB0
5mg Each for $200.11

AAJ66399MCR

 
alfa aesar™
J66399MCR
1mg Each for $68.35
Description & Specifications

Specifications

Product Type Dansyl-Tyr-Val-Gly
Molecular Weight 570.66
Storage Requirements -30° to -10°C
Sequence Dansyl-Y-V-G
Format Solid
Regulatory Status RUO
Purification Method HPLC