missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Danofloxacin

Synthetic fluoroquinone with broad spectrum anti-bacterial activity

$280.66 - $280.66

Chemical Identifiers

CAS 112398-08-0
Molecular Formula C19H20FN3O3
Molecular Weight (g/mol) 357.385
MDL Number MFCD00864910
InChI Key QMLVECGLEOSESV-RYUDHWBXSA-N
Synonym danofloxacin, danofloxacine, danofloxacino, danofloxacinum, unii-24cu1ys91d, dsstox_cid_26432, dsstox_rid_81609, dsstox_gsid_46432, danofloxacin inn:ban, danofloxacine inn-french
PubChem CID 71335
IUPAC Name 1-cyclopropyl-6-fluoro-7-[(1S,4S)-5-methyl-2,5-diazabicyclo[2.2.1]heptan-2-yl]-4-oxoquinoline-3-carboxylic acid
SMILES CN1CC2CC1CN2C3=C(C=C4C(=C3)N(C=C(C4=O)C(=O)O)C5CC5)F
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ62904ME View Documents Alfa Aesar™
J62904ME
500mg Each for $280.66
 
Description

Description

Solubility
Soluble in water, acetic acid and DMSO

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Specifications

Chemical Identifiers

112398-08-0
357.385
QMLVECGLEOSESV-RYUDHWBXSA-N
71335
CN1CC2CC1CN2C3=C(C=C4C(=C3)N(C=C(C4=O)C(=O)O)C5CC5)F
C19H20FN3O3
MFCD00864910
danofloxacin, danofloxacine, danofloxacino, danofloxacinum, unii-24cu1ys91d, dsstox_cid_26432, dsstox_rid_81609, dsstox_gsid_46432, danofloxacin inn:ban, danofloxacine inn-french
1-cyclopropyl-6-fluoro-7-[(1S,4S)-5-methyl-2,5-diazabicyclo[2.2.1]heptan-2-yl]-4-oxoquinoline-3-carboxylic acid

Specifications

White
500mg
C19H20FN3O3
14,2813
Soluble in water, acetic acid and DMSO
CN1CC2CC1CN2C3=C(C=C4C(=C3)N(C=C(C4=O)C(=O)O)C5CC5)F
357.385
357.38
Danofloxacin
268°C to 272°C
112398-08-0
MFCD00864910
danofloxacin, danofloxacine, danofloxacino, danofloxacinum, unii-24cu1ys91d, dsstox_cid_26432, dsstox_rid_81609, dsstox_gsid_46432, danofloxacin inn:ban, danofloxacine inn-french
QMLVECGLEOSESV-RYUDHWBXSA-N
1-cyclopropyl-6-fluoro-7-[(1S,4S)-5-methyl-2,5-diazabicyclo[2.2.1]heptan-2-yl]-4-oxoquinoline-3-carboxylic acid
71335
Powder