Please login to your online account to display your discounted pricing

Dacarbazine, Fisher BioReagents


Quantity 250mg
Packaging Amber Glass

Manufacturer: fisher scientific company  BP2513250

 View more versions of this product

Catalog No. BP2513250

  • / Each
  • / Case of 5 Each
 

Description & Specifications

Chemical Identifiers

Melting Point 202°C
Formula Weight 182.2
Physical Form Powder/Solid
Quantity 250mg
Merck Index 15, 2795
Molecular Formula C6H10N6O
Color Off-white
Synonym 5-[3, 3-Dimethyl-1-triazenyl]imidazole-4-carboxamide; DTIC
Chemical Name or Material Dacarbazine
CAS 4342-03-4
Recommended Storage 4° C
Identification Pass Test
Packaging Amber Glass
ChemAlert Storage Symbol Gray

Specifications

Chemical Name or Material Dacarbazine
CAS 4342-03-4
Physical Form Powder/Solid
Molecular Formula C6H10N6O
Formula Weight 182.2
Synonym 5-[3, 3-Dimethyl-1-triazenyl]imidazole-4-carboxamide; DTIC
Accessories
 

4° C

SDS & Safety Documents