Learn More
Dacarbazine, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals J61023ME
Description
Used in treatment of malignant melanoma and sarcomas. Generates methyl adducts to purine bases in DNA
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Used in treatment of malignant melanoma and sarcomas
- Generates methyl adducts to purine bases in DNA
Specifications
Dacarbazine | |
210°C to 212°C | |
C4H3IN2O | |
500 mg | |
5-iodopyrimidin-4-ol, 5-iodopyrimidin-4 3h-one, 4-hydroxy-5-iodopyrimidine, 5-iodo-1h-pyrimidin-4-one, 4 1h-pyrimidinone, 5-iodo, 5-iodopyrimidin-4 1h-one, 5-iodo-3h-pyrimidin-4-one, 5-iodo-3,4-dihydropyrimidin-4-one, 5-iodopyrimidin-4-ol;5-iodopyrimidin-4 1h-one | |
SMIGOGPUTQXOKF-UHFFFAOYSA-N | |
5-iodo-1H-pyrimidin-6-one | |
12356582 | |
Powder |
4342-03-4 | |
White to Yellow | |
MFCD00057167 | |
14,2798 | |
Slightly soluble in water,DMSO and ethanol /n | |
C1=C(C(=O)NC=N1)I | |
221.99 | |
182.18 |
Chemical Identifiers
4342-03-4 | |
MFCD00057167 |
Safety and Handling
GHS H Statement
H311-H340-H350-H332-H315-H319-H335
Toxic in contact with skin.
May cause genetic defects.
May cause cancer.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P332+P313-P362-P501c
H312+H332-H315-H319-H335-H350
EINECSNumber : 224-396-1
RTECSNumber : NI3950000
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only