Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ D4-GDI, Rabbit Polyclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry and western blotting experiments with Thermo Scientific™ D4-GDI, Rabbit Polyclonal Antibody.

$726.14 - $1,213.55

Specifications

Antigen D4-GDI
Applications Western Blotting
Classification Polyclonal
Conjugate Unlabeled
Cross Reactivity Human
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

RB9042P

 
thermo scientific™ lab vision™
RB9042P
1mL Each for $1,213.55

RB9042P1

 
thermo scientific™ lab vision™
RB9042P1
500µL Each for $726.82

RB9042R7

 
thermo scientific™ lab vision™
RB9042R7
7mL Each for $726.14
Description & Specifications

Specifications

Antigen D4-GDI
Applications Western Blotting
Classification Polyclonal
Conjugate Unlabeled
Cross Reactivity Human
Host Species Rabbit
Regulatory Status RUO
Research Discipline Apoptosis
Type Primary
Immunogen Recombinant full-length human D4-GDI protein
Isotype IgG

Obtain accurate, reproducible results in immunohistochemistry and western blotting experiments with Thermo Scientific™ D4-GDI, Rabbit Polyclonal Antibody.

D4-GDI belongs to the family of Rho-GDIs or GDP-dissociation inhibitors for the Rho/Rac family of small G proteins; consisting of RhoGDI-1, D4-GDI/RhoGDI-2 and RhoGDI-3. It inhibits the release of GDP from the Rho proteins, essential for GTP-loading and activation of GTPase activity. D4-GDI is located in the cytoplasm and in involved in the regulation of the membrane association and dissociation cycle of Rho/Rac proteins. It is known to be a substrate for caspase 3 during Fas-mediated apoptosis.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human. Others not tested.

Epitope: Not determined

Immunogen: Recombinant full-length human D4-GDI protein

Molecular Weight: 28 kDa

Positive Control: Placenta and tonsil

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO