Please login to your online account to display your discounted pricing

Alfa Aesar™ D-Valine methyl ester hydrochloride, 99%

$54.19 - $172.41

Chemical Identifiers

Chemical Name or Material D-Valine methyl ester hydrochloride
CAS 7146-15-8
Molecular Formula C6H13NO2·HCl
Formula Weight 167.64
MDL Number MFCD00237309
Beilstein 3912091
Synonym H-D-Val-Ome·HCl
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2688303

  SDS
alfa aesar™
H2688303
1g Each for $54.19

AAH2688306

  SDS
alfa aesar™
H2688306
5g Each for $172.41
Description & Specifications

Specifications

Formula Weight 167.64
Chemical Name or Material D-Valine methyl ester hydrochloride
Assay 99%
Molecular Formula C6H13NO2·HCl
MDL Number MFCD00237309
CAS 7146-15-8
Beilstein 3912091
Synonym H-D-Val-Ome·HCl
Melting Point ≈170°C
Quantity 1g
Recommended Storage 2° to 8°C
Safety and Handling

2° to 8°C