Please login to your online account to display your discounted pricing

Alfa Aesar™ [D-Val&superscript_2;&superscript_2;, Phe&superscript_3;&superscript_3;] Big Endothelin-1 (16-38), Human

For cellular and molecular biology applications

Specifications

Product Type [D-Val22, Phe33] Big Endothelin-1 (16-38), human
Cross Reactivity Human
Molecular Weight 2620.91
Storage Requirements -30° to -10°C
Sequence His-Leu-Asp-Ile-Ile-Trp-DVal-Asn-Thr-Pro-Glu-His-Val-Val-Pro-Tyr-Gly-Leu-Gly-Ser-Pro-Arg-Ser
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66758EXD

 
alfa aesar™
J66758EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66758MCR

 
alfa aesar™
J66758MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type [D-Val22, Phe33] Big Endothelin-1 (16-38), human
Cross Reactivity Human
Molecular Weight 2620.91
Storage Requirements -30° to -10°C
Sequence His-Leu-Asp-Ile-Ile-Trp-DVal-Asn-Thr-Pro-Glu-His-Val-Val-Pro-Tyr-Gly-Leu-Gly-Ser-Pro-Arg-Ser
Format Solid
Regulatory Status RUO
Purification Method HPLC