missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ D-Val-Leu-Arg-p-nitroanilide

For cellular and molecular biology applications

$117.32 - $421.08

Specifications

Product Type D-Val-Leu-Arg-p-nitroanilide
Molecular Weight 506.59
Storage Requirements -30° to -10°C
Sequence D-Val-Leu-Arg-pNA
Format Solid
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ66439LB0
View Documents
Alfa Aesar™
J66439LB0
5mg Each for $117.32
 
AAJ66439MB
View Documents
Alfa Aesar™
J66439MB
25mg Each for $421.08
 
Specifications

Specifications

D-Val-Leu-Arg-p-nitroanilide
-30° to -10°C
Solid
HPLC
506.59
D-Val-Leu-Arg-pNA
RUO

For Research Use Only