missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ D-Tyrosine ethyl ester hydrochloride, 98%

$50.43 - $220.53

Chemical Identifiers

CAS 23234-43-7
Molecular Formula C11H16ClNO3
Molecular Weight (g/mol) 245.703
MDL Number MFCD00180750
InChI Key BQULAXAVRFIAHN-HNCPQSOCSA-N
Synonym h-d-tyr-oet.hcl, h-d-tyr-oet hcl, d-tyrosine ethyl ester hydrochloride, r-ethyl 2-amino-3-4-hydroxyphenyl propanoate hydrochloride, ethyl d-tyrosinate hcl, ethyl d-tyrosinate hydrochloride, ethyl 2r-2-amino-3-4-hydroxyphenyl propanoate hydrochloride, h-d-tyr-oethcl, h-d-tyr-oet?cl, h-d-tyr-oet. hcl
PubChem CID 45357981
IUPAC Name ethyl (2R)-2-amino-3-(4-hydroxyphenyl)propanoate;hydrochloride
SMILES CCOC(=O)C(CC1=CC=C(C=C1)O)N.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6219406 View Documents Alfa Aesar™
H6219406
5g Each for $50.43
 
AAH6219414 View Documents Alfa Aesar™
H6219414
25g Each for $220.53
 
Specifications

Chemical Identifiers

23234-43-7
245.703
BQULAXAVRFIAHN-HNCPQSOCSA-N
45357981
CCOC(=O)C(CC1=CC=C(C=C1)O)N.Cl
C11H16ClNO3
MFCD00180750
h-d-tyr-oet.hcl, h-d-tyr-oet hcl, d-tyrosine ethyl ester hydrochloride, r-ethyl 2-amino-3-4-hydroxyphenyl propanoate hydrochloride, ethyl d-tyrosinate hcl, ethyl d-tyrosinate hydrochloride, ethyl 2r-2-amino-3-4-hydroxyphenyl propanoate hydrochloride, h-d-tyr-oethcl, h-d-tyr-oet?cl, h-d-tyr-oet. hcl
ethyl (2R)-2-amino-3-(4-hydroxyphenyl)propanoate;hydrochloride

Specifications

23234-43-7
MFCD00180750
BQULAXAVRFIAHN-HNCPQSOCSA-N
ethyl (2R)-2-amino-3-(4-hydroxyphenyl)propanoate;hydrochloride
45357981
98%
D-Tyrosine ethyl ester hydrochloride
C11H16ClNO3
h-d-tyr-oet.hcl, h-d-tyr-oet hcl, d-tyrosine ethyl ester hydrochloride, r-ethyl 2-amino-3-4-hydroxyphenyl propanoate hydrochloride, ethyl d-tyrosinate hcl, ethyl d-tyrosinate hydrochloride, ethyl 2r-2-amino-3-4-hydroxyphenyl propanoate hydrochloride, h-d-tyr-oethcl, h-d-tyr-oet?cl, h-d-tyr-oet. hcl
CCOC(=O)C(CC1=CC=C(C=C1)O)N.Cl
245.703
245.7
5g