Please login to your online account to display your discounted pricing

Alfa Aesar™ D-Tyr-Val-Gly

$80.47 - $80.47

Chemical Identifiers

Chemical Name or Material D-Tyr-Val-Gly
CAS 86030-52-6
Molecular Formula C16H23N3O5
Formula Weight 337.4
MDL Number MFCD00063349
Synonym dY-V-G
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66654MCR

 
alfa aesar™
J66654MCR
1mg Each for $80.47
Description & Specifications

Specifications

Formula Weight 337.4
Chemical Name or Material D-Tyr-Val-Gly
MDL Number MFCD00063349
Physical Form Solid
Molecular Formula C16H23N3O5
Quantity 1mg
Synonym dY-V-G
Recommended Storage −30° to −10°C
Storage Note 1 -30° to -10°C
Color White to off-white
CAS 86030-52-6
Safety and Handling

−30° to −10°C

-30° to -10°C