Please login to your online account to display your discounted pricing

Alfa Aesar™ D-Tryptophan ethyl ester hydrochloride, 98%

Chemical Identifiers

Chemical Name or Material D-Tryptophan ethyl ester hydrochloride
CAS 61535-49-7
Formula Weight 268.74
MDL Number MFCD00067552
Synonym H-D-Trp-OEt.HCl; 51535-49-1 Not valid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6692903

 
alfa aesar™
H6692903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6692906

 
alfa aesar™
H6692906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 268.74
Assay 98%
MDL Number MFCD00067552
CAS 61535-49-7
Synonym H-D-Trp-OEt.HCl; 51535-49-1 Not valid
Physical Form Solid
Chemical Name or Material D-Tryptophan ethyl ester hydrochloride
Quantity 1g