missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

D(+)-Tryptophan, 99%, ACROS Organics™

Manufacturer:  Acros Organics AC168635000

 View more versions of this product

Catalog No. AC168635000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

D(+)-Tryptophan
(1% aq. soln.) Clear colorless to light yellow
Authentic
0.5% max.
QIVBCDIJIAJPQS-SECBINFHSA-N
(2R)-2-amino-3-(1H-indol-3-yl)propanoic acid
9060
CHEBI:16296
204.23
99%
White to Yellow
500g
153-94-6
99%
C11H12N2O2
d-tryptophan, h-d-trp-oh, r-tryptophan, d +-tryptophan, d-trytophane, d-tryptophane, tryptophan, d, 2r-2-amino-3-1h-indol-3-yl propanoic acid, +-tryptophan, h-d-typ-oh
C1=CC=C2C(=C1)C(=CN2)CC(C(=O)O)N
204.229
+ 31.50
Solubility in water: 11g/L (20°C). Other solubilities: soluble in alkali hydroxides, soluble in hot alcohol, insoluble in chloroform
Powder
+ 31.50 (24.00°C c=1,H2O)
282.0°C to 285.0°C

Chemical Identifiers

153-94-6
204.229
d-tryptophan, h-d-trp-oh, r-tryptophan, d +-tryptophan, d-trytophane, d-tryptophane, tryptophan, d, 2r-2-amino-3-1h-indol-3-yl propanoic acid, +-tryptophan, h-d-typ-oh
CHEBI:16296
C1=CC=C2C(=C1)C(=CN2)CC(C(=O)O)N
C11H12N2O2
QIVBCDIJIAJPQS-SECBINFHSA-N
9060
(2R)-2-amino-3-(1H-indol-3-yl)propanoic acid
SDS Documents