missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

D-(+)-Trehalose, Dihydrate, Fisher BioReagents

Manufacturer:  Fisher Scientific Company BP268725

 View more versions of this product

Catalog No. BP268725



 

Specifications Description & Specifications

Specifications

D-(+)-Trehalose, Dihydrate
Pass Test
Conforms to reference
98% min.
≥98 %
378.33
97°C
25g
6138-23-4
8.5 to 10.5%
Poly Bottle
Powder/Solid
C12H22O11 · 2 H2O
15, 9742
Off-white

Chemical Identifiers

6138-23-4
≥98 %
378.33
Powder/Solid
C12H22O11 · 2 H2O
D-(+)-Trehalose, Dihydrate

Safety and Handling

SDS & Safety Documents

WARNING!

Emergency Overview
Irritating to eyes and skin. Wear personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:2
Flammability:1
Instability:1

Recommended Storage : RT