Learn More
Thermo Scientific Chemicals D-Trehalose, 99%, anhydrous
$221.91 - $2137.42
Chemical Identifiers
CAS | 99-20-7 |
---|---|
Molecular Formula | C12H22O11 |
Molecular Weight (g/mol) | 342.30 |
MDL Number | MFCD00006628 |
InChI Key | HDTRYLNUVZCQOY-LBYRQFQKNA-N |
Synonym | trehalose, d-trehalose, alpha,alpha-trehalose, mycose, ergot sugar, alpha-d-trehalose, d-+-trehalose, d-+-trehalose, anhydrous, alpha,alpha'-trehalose, trehaose |
PubChem CID | 7427 |
ChEBI | CHEBI:16551 |
IUPAC Name | (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxane-3,4,5-triol |
SMILES | OC[C@H]1O[C@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC309870250
|
Thermo Scientific Chemicals
309870250 |
25 g | Plastic bottle |
Each for $221.91
|
|
||||
AC309871000
|
Thermo Scientific Chemicals
309871000 |
100 g | Plastic bottle |
Each for $612.47
|
|
||||
AC309875000
|
Thermo Scientific Chemicals
309875000 |
500 g | Plastic bottle |
Each for $2,137.42
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
99-20-7 | |
342.30 | |
HDTRYLNUVZCQOY-LBYRQFQKNA-N | |
7427 | |
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxane-3,4,5-triol |
C12H22O11 | |
MFCD00006628 | |
trehalose, d-trehalose, alpha,alpha-trehalose, mycose, ergot sugar, alpha-d-trehalose, d-+-trehalose, d-+-trehalose, anhydrous, alpha,alpha'-trehalose, trehaose | |
CHEBI:16551 | |
OC[C@H]1O[C@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O |
Specifications
99-20-7 | |
White | |
98.5% min. (ELSD) on dry basis (HPLC) | |
C12H22O11 | |
25 g | |
+197.50 (20.00°C c=2,H2O) | |
trehalose, d-trehalose, alpha,alpha-trehalose, mycose, ergot sugar, alpha-d-trehalose, d-+-trehalose, d-+-trehalose, anhydrous, alpha,alpha'-trehalose, trehaose | |
HDTRYLNUVZCQOY-LBYRQFQKNA-N | |
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxane-3,4,5-triol | |
7427 | |
342.3 | |
Crystalline Powder |
203.0°C | |
Authentic | |
Plastic bottle | |
MFCD00006628 | |
15,9742 | |
+ 197.50 | |
(10% in water) Clear colorless to light yellow | |
OC[C@H]1O[C@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O | |
342.30 | |
CHEBI:16551 | |
99% | |
D-Trehalose, Anhydrous |
Safety and Handling
GHS H Statement:
Causes skin irritation.
GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber : 202-739-6
RUO – Research Use Only