missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ D-Tagatose, 99%

$56.51 - $699.49

Chemical Identifiers

CAS 87-81-0
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.156
MDL Number MFCD00134449
InChI Key BJHIKXHVCXFQLS-PQLUHFTBSA-N
Synonym 3s,4s,5r-1,3,4,5,6-pentahydroxyhexan-2-one, d--tagatose, d-lyxo-hex-2-ulose, keto-d-tagatose, lyxo-2-hexulose, naturlose, tagatose nf, tagatose, d, unii-t7a20y888y
PubChem CID 92092
ChEBI CHEBI:47693
IUPAC Name (3S,4S,5R)-1,3,4,5,6-pentahydroxyhexan-2-one
SMILES C(C(C(C(C(=O)CO)O)O)O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2119201
View Documents
Thermo Scientific™
B2119201
0.25g
Each for $56.51
 
AAB2119203
View Documents
Thermo Scientific™
B2119203
1g
Each for $176.03
 
AAB2119206
View Documents
Thermo Scientific™
B2119206
5g
Each for $699.49
 
Description

Description

Specifications

Chemical Identifiers

87-81-0
180.156
BJHIKXHVCXFQLS-PQLUHFTBSA-N
92092
(3S,4S,5R)-1,3,4,5,6-pentahydroxyhexan-2-one
C6H12O6
MFCD00134449
3s,4s,5r-1,3,4,5,6-pentahydroxyhexan-2-one, d--tagatose, d-lyxo-hex-2-ulose, keto-d-tagatose, lyxo-2-hexulose, naturlose, tagatose nf, tagatose, d, unii-t7a20y888y
CHEBI:47693
C(C(C(C(C(=O)CO)O)O)O)O

Specifications

−4.7° (c=1 in Water)
0.25g
C6H12O6
1724555
3s,4s,5r-1,3,4,5,6-pentahydroxyhexan-2-one, d--tagatose, d-lyxo-hex-2-ulose, keto-d-tagatose, lyxo-2-hexulose, naturlose, tagatose nf, tagatose, d, unii-t7a20y888y
C(C(C(C(C(=O)CO)O)O)O)O
180.156
CHEBI:47693
99%
129°C to 135°C
87-81-0
MFCD00134449
14,9030
BJHIKXHVCXFQLS-PQLUHFTBSA-N
(3S,4S,5R)-1,3,4,5,6-pentahydroxyhexan-2-one
92092
180.16
D-Tagatose
Safety and Handling

Safety and Handling

EINECSNumber : 201-772-3

TSCA : No

SDS
Product Certifications

RUO – Research Use Only