Please login to your online account to display your discounted pricing

D(+)-Sucrose, ACS reagent, ACROS Organics™


Quantity 25kg

Manufacturer: thermo fisher scientific  AC424500250

 View more versions of this product

Catalog No. AC424500250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Purity Grade ACS Reagent Grade
Formula Weight 342.29
Physical Form Crystals
Quantity 25kg
Color White
Specific Rotation +66.3° to +66.8° (25°C, 589nm) (c=26, H2O)
Iron (Fe) 5ppm max.
Titratable Acid 0.0008 meq/g max.
Chlorine (Cl) 50ppm max.
Synonym Saccharose
CAS 57-50-1
Insoluble Matter 0.005% max.
Residue after Ignition 0.01% max.
Loss on Drying 0.03% max. (105°C)
Invert Sugars 0.05% max.
Heavy Metals (as Pb) 5ppm max.
Sulfate and Sulfite 0.005% max.
Infrared Spectrum Authentic
MDL Number MFCD00006626
Chemical Name or Material D(+)-Sucrose
Molecular Formula C12H22O11

Specifications

Chemical Name or Material D(+)-Sucrose
CAS 57-50-1
Purity Grade ACS Reagent Grade
Molecular Formula C12H22O11
Formula Weight 342.29
MDL Number MFCD00006626
Synonym Saccharose