Please login to your online account to display your discounted pricing

D(+)-Sucrose, 99.7%, for biochemistry, ACROS Organics™

Chemical Identifiers

Chemical Name or Material D(+)-Sucrose
CAS 57-50-1
Purity Grade For biochemistry
Assay ≥99.7% (HPLC)
Assay Percent Range 99.7%
Molecular Formula C12H22O11
Formula Weight 342.29
MDL Number MFCD00006626
Synonym Saccharose
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC177140250

  SDS
thermo fisher scientific
AC177140250
Plastic bag in resealable cardbox 25kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥99.7% (HPLC)
Formula Weight 342.29
Physical Form Liquid
Quantity 25kg
Color Clear colorless
Purity Grade For biochemistry
pH 6.5 to 7.5 (10% soln.)
Water 0.06% max.
Loss on Drying 0.1 % max.
Infrared Spectrum Authentic
Packaging Plastic bag in resealable cardbox
Sulfated Ash 0.02 % max.
Reducing Sugars 0.04 % max.
Iron (Fe) 0.5ppm max.
Specific Rotation (20°C, 589nm) (c=26, H2O) > +65.9°
Insoluble in Water 0.0007 % max.
MDL Number MFCD00006626
Molecular Formula C12H22O11
Assay Percent Range 99.7%
CAS 57-50-1
Sulfite 15ppm (as SO2) max.
Heavy Metals (as Pb) 5ppm max.
Trace Analysis 15ppm max.
Turbidity 3 NTU (in water) max.
Synonym Saccharose
Chemical Name or Material D(+)-Sucrose